Methotrexate

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioic acid

J63075-03 J63075-MC
CAAAJ63075-03EA 1123.38 CAD
CAAAJ63075-03 CAAAJ63075-MC
Methotrexate
Methotrexate

Anti-inflammatory, cytotoxic drug that acts as a potent folic acid antagonist.


Powder

Formula: C₂₀H₂₂N₈O₅
MW: 454.44 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00150847
CAS Number: 59-05-2
EINECS: 200-413-8
UN: 1544
ADR: 6.1 III
Merck Index: 14,5985

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR