Tyrphostin A9 ≥99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
[4-Hydroxy-3,5-bis(2-methyl-2-propanyl)benzylidene]malononitrile, Malonoben
J63058-MC
J63058-MF
CAAAJ63058-MCEA
372.88
CAD
CAAAJ63058-MC
CAAAJ63058-MF
Tyrphostin A9 ≥99%
Tyrphostin A9
Selective inhibitor of PDGF receptor tyrosine kinase
Powder
Applications: Selective inhibitor of PDGF receptor tyrosine kinase
Formula:
C₁₈H₂₂N₂O Storage Temperature: Ambient |
MDL Number:
MFCD00209853 CAS Number: 10537-47-0 UN: 2811 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...