Arsenazo III

Supplier: Thermo Scientific Chemicals
Danger

J63031-06 J63031-14
CAAAJ63031-06EA 369.72 CAD
CAAAJ63031-06 CAAAJ63031-14
Arsenazo III
Arsenazo III

Suitable for determination of micromolar amounts of calcium


Powder

Formula: C₂₂H₁₈As₂N₄O₁₄S₂
Storage Temperature: Ambient
MDL Number: MFCD00036695
CAS Number: 1668-00-4
EINECS: 216-788-6
UN: 3465
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR