α-Terpinene 90+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: p-Mentha-1,3-diene, 1-Isopropyl-4-methylcyclohexa-1,3-diene

J62871-AE J62871-AP
CAAAJ62871-AEEA 51.98 CAD
CAAAJ62871-AE CAAAJ62871-AP
α-Terpinene 90+%
α-Terpinene

Liquid


CAS: 99-86-5 MDL: MFCD00001534

Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 173
Melting Pt: -48
Density: 0.837
Flash Pt: 50 °C (122 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001534
CAS Number: 99-86-5
EINECS: 202-795-1
UN: 2319
ADR: 3 III
Merck Index: 14,9170

Order Now

Specification Test Results

Assay Minimum 90


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR