Bupivacaine hydrochloride monohydrate ≥98%

Supplier: Thermo Scientific Chemicals
Danger

J62835-03 J62835-06 J62835-14
CAAAJ62835-03EA 104.75 CAD
CAAAJ62835-03 CAAAJ62835-06 CAAAJ62835-14
Bupivacaine hydrochloride monohydrate ≥98%
Bupivacaine hydrochloride monohydrate

Inhibits TREK-1 channels and depolarizes the cell membrane


White powder


Applications: Inhibits TREK-1 channels and depolarizes the cell membrane

Formula: C₁₈H₃₁ClN₂O₂
Melting Pt: 255…259 °C
Storage Temperature: Ambient
MDL Number: MFCD00078956
CAS Number: 73360-54-0
EINECS: 241-917-8
UN: 2811
ADR: "6.1",III
Merck Index: 14,1495

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR