Deblock trichloroacetic acid solution 3% (w/w) in dichloromethane

Supplier: Thermo Scientific Chemicals
Danger

J62821-AO J62821-K4
CAAAJ62821-AHEA 107.33 CAD
CAAAJ62821-AH CAAAJ62821-AO CAAAJ62821-K4
Deblock trichloroacetic acid solution 3% (w/w) in dichloromethane
Deblock trichloroacetic acid solution

Solvent specially purified and analyzed for biotechnology applications, including peptide and oligonucleotide synthesis


MDL: MFCD00004177

Density: 1.325
Storage Temperature: Ambient
MDL Number: MFCD00004177
EINECS: 200-927-2
UN: 2922
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR