Ivermectin

Supplier: Thermo Scientific Chemicals
Danger

J62777-03 J62777-MD
CAAAJ62777-03EA 134.93 CAD
CAAAJ62777-03 CAAAJ62777-MD
Ivermectin
Ivermectin

Antiparasitic and macrolide antibiotic. Modulates glutamate-GABA-activated chloride channels. Also activates the glycine receptor chloride channel


Powder


Applications: Antiparasitic and macrolide antibiotic. Modulates glutamate-GABA-activated chloride channels. Also activates the glycine receptor chloride channel

Formula: C₄₈H₇₄O₁₄
Storage Temperature: Ambient
MDL Number: MFCD00869511
CAS Number: 70288-86-7
EINECS: 274-536-0
UN: 2811
ADR: "6.1",III
Merck Index: 14,5248

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR