Ketoprofen
Supplier: Thermo Scientific Chemicals
|
Danger
|
J62702-06
J62702-14
CAAAJ62702-06EA
146.15
CAD
CAAAJ62702-06
CAAAJ62702-14
Ketoprofen
Ketoprofen
A non-steroidal anti-inflammatory drug (NSAID); also inhibits cyclooxygenases-1 and -2
Powder
Formula:
C₁₆H₁₄O₃ Melting Pt: 92…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055790 CAS Number: 22071-15-4 EINECS: 244-759-8 UN: 2811 ADR: "6.1",III Merck Index: 14,5305 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...