Milrinone ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
J62659-MA
J62659-MF
CAAAJ62659-MAEA
274.92
CAD
CAAAJ62659-MA
CAAAJ62659-MC
CAAAJ62659-MF
Milrinone ≥98%
Milrinone
Potent cAMP-specific phosphodiesterase inhibitor
Powder
Applications: Potent cAMP-specific phosphodiesterase inhibitor
Formula:
C₁₂H₉N₃O Melting Pt: 264 °C Storage Temperature: Ambient |
MDL Number:
MFCD00133539 CAS Number: 78415-72-2 EINECS: 278-903-6 UN: 3439 ADR: 6.1,III Merck Index: 14,6197 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...