Amrinone 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
J62655-03
J62655-MD
CAAAJ62655-03EA
360.24
CAD
CAAAJ62655-03
CAAAJ62655-MD
Amrinone 98%
Amrinone
Phosphodiesterase III inhibitor and inhibits platelet aggregation
Crystals
Applications: Phosphodiesterase III inhibitor and inhibits platelet aggregation
Formula:
C₁₀H₉N₃O Melting Pt: 294…297 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00083228 CAS Number: 60719-84-8 EINECS: 262-390-0 UN: 2811 ADR: 6.1,III Merck Index: 14,592 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...