Sorbitan sesquioleate

Supplier: Thermo Scientific Chemicals

J62648-AP J62648-K2
CAAAJ62648-APEA 77.89 CAD
CAAAJ62648-AP CAAAJ62648-K2
Sorbitan sesquioleate
Sorbitan sesquioleate

An emulsifier


Viscous liquid


Applications: An emulsifier

Formula: C₄₂H₇₆O₇
Density: 0.989
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00151568
CAS Number: 8007-43-0
EINECS: 232-360-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR