Erythrosin B, powder, spirit soluble
Supplier: Thermo Scientific Chemicals
J62619-06
J62619-14
J62619-22
CAAAJ62619-06EA
62.66
CAD
CAAAJ62619-06
CAAAJ62619-14
CAAAJ62619-22
Erythrosin B, powder, spirit soluble
Erythrosin B
CAS: 15905-32-5
MDL: MFCD00005044
Formula:
C₂₀H₈I₄O₅ Storage Temperature: Ambient |
MDL Number:
MFCD00005044 CAS Number: 15905-32-5 EINECS: 240-046-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...