Diclofenac sodium salt
Supplier: Thermo Scientific Chemicals
|
Danger
|
J62609-06
J62609-14
J62609-22
CAAAJ62609-06EA
63.04
CAD
CAAAJ62609-06
CAAAJ62609-14
CAAAJ62609-22
Diclofenac sodium salt
Diclofenac sodium salt
Standard NSAID and cyclooxygenase (COX) inhibitor.
Powder
Formula:
C₁₄H₁₀Cl₂NNaO₂ Melting Pt: 283…285 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082251 CAS Number: 15307-79-6 EINECS: 239-346-4 UN: 2811 ADR: "6.1",III Merck Index: 14,3081 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...