Doxepin hydrochloride

Supplier: Thermo Scientific Chemicals
Danger

J62579-06 J62579-14
CAAAJ62579-06EA 229.1 CAD
CAAAJ62579-06 CAAAJ62579-14
Doxepin hydrochloride
Doxepin hydrochloride

Potent H1-histamine receptor antagonist and a 5-HT receptor antagonist


Powder


Applications: Potent H1-histamine receptor antagonist and a 5-HT receptor antagonist

Formula: C₁₉H₂₂ClNO
Melting Pt: 187…189 °C
Storage Temperature: Ambient
MDL Number: MFCD00079135
CAS Number: 1229-29-4
EINECS: 214-966-8
UN: 2811
ADR: "6.1",III
Merck Index: 14,3435

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR