Acrylamide 40% in aqueous solution bisacrylamide-free

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Prop-2-enamide

J62480-AP J62480-K2
CAAAJ62480-APEA 234.35 CAD
CAAAJ62480-AP CAAAJ62480-K2
Acrylamide 40% in aqueous solution bisacrylamide-free
Acrylamide (monomer)

Liquid


CAS: 79-06-1 MDL: MFCD00008032

Formula: C₃H₅NO
Storage Temperature: Ambient
MDL Number: MFCD00008032
CAS Number: 79-06-1
EINECS: 201-173-7
UN: 3426
ADR: 6.1,III
Merck Index: 14,129
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7687b7f17a8cdf31c559a33fd5d4021a2e50a40a&vid=fecebe0c34b037788df119c11bfc9637d78cda0d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR