Brefeldin A 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(1R,2E,6S,10E,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-6,7,8,9,12,13,14,14a-octahydro-1H-cyclopenta[f][1]oxacyclotridecin-4(11aH)-one, Brefeldin A, natural
J62340-MA
J62340-MB
J62340-MF
CAAAJ62340-MAEA
224.36
CAD
CAAAJ62340-MA
CAAAJ62340-MB
CAAAJ62340-MF
Brefeldin A 99%
Brefeldin A
Inhibits protein transport from the endoplasmic reticulum to Golgi apparatus. Brefeldin A mediated apoptosis has been observed in human tumor cells.
Crystalline powder
Formula:
C₁₆H₂₄O₄ Melting Pt: 201…205 °C Storage Temperature: Freezer |
MDL Number:
MFCD00083258 CAS Number: 20350-15-6 Merck Index: 14,1369 |
Specification Test Results
Assay Minimum | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...