Neostigmine bromide

Supplier: Thermo Scientific Chemicals
Danger

J62300-03 J62300-MD
CAAAJ62300-03EA 153.1 CAD
CAAAJ62300-03 CAAAJ62300-MD
Neostigmine bromide
Neostigmine bromide

A reversible acetylcholinesterase inhibitor


Powder


Applications: A reversible acetylcholinesterase inhibitor

Formula: C₁₂H₁₉BrN₂O₂
Melting Pt: 175…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00011795
CAS Number: 114-80-7
EINECS: 204-054-8
UN: 2811
ADR: "6.1",III
Merck Index: 14,6464

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR