Amlodipine ≥97%

Supplier: Thermo Scientific Chemicals
Danger

J62242-03 J62242-06 J62242-09
CAAAJ62242-03EA 221.2 CAD
CAAAJ62242-03 CAAAJ62242-06 CAAAJ62242-09
Amlodipine ≥97%
Amlodipine

A calcium channel antagonist with potent antioxidant activity


Crystalline powder


Applications: A calcium channel antagonist with potent antioxidant activity

Formula: C₂₀H₂₅ClN₂O₅
Melting Pt: 134…136 °C
Storage Temperature: Ambient
MDL Number: MFCD00864687
CAS Number: 88150-42-9
EINECS: 425-820-1
UN: 2811
ADR: "6.1",III
Merck Index: 14,491

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR