Amiloride hydrochloride dihydrate

Supplier: Thermo Scientific Chemicals
Danger

J62168-03 J62168-06
CAAAJ62168-03EA 97.96 CAD
CAAAJ62168-03 CAAAJ62168-06
Amiloride hydrochloride dihydrate
Amiloride hydrochloride dihydrate

Sodium channel blocker


Powder


Applications: Sodium channel blocker

Formula: C₆H₁₃Cl₂N₇O₃
Melting Pt: >240 °C
Storage Temperature: Ambient
MDL Number: MFCD00058197
CAS Number: 17440-83-4
EINECS: 217-958-2
UN: 2811
ADR: "6.1",III
Merck Index: 14,406

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR