Ergocalciferol (vitamin D2, calciferol) 98%

Supplier: Thermo Scientific Chemicals
Danger

J62163-03 J62163-06
CAAAJ62163-03EA 65.57 CAD
CAAAJ62163-03 CAAAJ62163-06
Ergocalciferol (vitamin D2, calciferol) 98%
Ergocalciferol (Vitamin D2, Calciferol)

Powder


CAS: 50-14-6 MDL: MFCD00166988

Formula: C₂₈H₄₄O
Melting Pt: 118…119 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00166988
CAS Number: 50-14-6
EINECS: 200-014-9
UN: 2811
ADR: "6.1",III
Merck Index: 14,10018

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR