Ethidium bromide 0.625 mg/ml in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
, 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide
J62029-AA
J62029-AA
CAAAJ62029-AAEA
117.08
CAD
CAAAJ62029-AA
AAJ62029-AA
Ethidium bromide 0.625 mg/ml in aqueous solution
Ethidium bromide
For visualization of DNA in gel electrophoresis
Formula:
C₂₁H₂₀BrN₃ MW: 394.31 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011724 CAS Number: 1239-45-8 UN: 2810 ADR: "6.1",III Merck Index: 14,4731 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...