Phenoxybenzamine hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

J61900-03 J61900-MD
CAAAJ61900-03EA 76 CAD
CAAAJ61900-03 CAAAJ61900-MD
Phenoxybenzamine hydrochloride
Phenoxybenzamine hydrochloride

Selective alpha adrenergic blocking agent. Irreversible calmodulin inhibitor


Powder


Applications: Selective alpha adrenergic blocking agent. Irreversible calmodulin inhibitor

Formula: C₁₈H₂₂ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD00055152
CAS Number: 63-92-3
EINECS: 200-569-7
Merck Index: 14,7256

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR