Ammonium persulfate (APS) for electrophoresis
Supplier: Thermo Scientific Chemicals
|
Danger
|
J61856-14
J61856-22
CAAAJ61856-14EA
66.36
CAD
CAAAJ61856-14
CAAAJ61856-22
Ammonium persulfate (APS) for electrophoresis
Ammonium peroxodisulphate (APS)
Powder
CAS: 7727-54-0
MDL: MFCD00003390
Formula:
(NH₄)₂S₂O₈ Melting Pt: 120 °C (dec.) Density: 1.982 Storage Temperature: Ambient |
MDL Number:
MFCD00003390 CAS Number: 7727-54-0 EINECS: 231-786-5 UN: 1444 ADR: 5.1,III Merck Index: 14,541 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...