Thiomersal
Supplier: Thermo Scientific Chemicals
|
Danger
|
J61799-09
J61799-14
CAAAJ61799-09EA
217.27
CAD
CAAAJ61799-09
CAAAJ61799-14
Thiomersal
Thiomersal
A bacteriostatic and fungistatic mercurial agent
Powder
Formula:
2-(C₂H₅HgS)C₆H₄CO₂Na Melting Pt: 234…237 °C Flash Pt: 250 °C (482 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013062 CAS Number: 54-64-8 EINECS: 200-210-4 UN: 2025 ADR: 6.1,III Merck Index: 14,9317 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...