Histamine (free base)

Supplier: Thermo Scientific Chemicals
Danger

J61727-03 J61727-06
CAAAJ61727-03EA 159.58 CAD
CAAAJ61727-03 CAAAJ61727-06
Histamine (free base)
Histamine (free base)

Endogenous H1 and H2 histamine receptor agonist; H1 activation mobilizes calcium; H2 activation stimulates adenylate cyclase activity in neurons; activates nitric oxide synthetase; potent vasodilator


Powder

Formula: C₅H₉N₃
Storage Temperature: Refrigerator
MDL Number: MFCD00005210
CAS Number: 51-45-6
EINECS: 200-100-6
UN: 2811
ADR: 6.1,III
Merck Index: 14,4719

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR