Clozapine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 8-Chloro-11-(4-methyl-1-piperazinyl)-5H-dibenzo[b,e][1,4]diazepine

J61583-MB J61583-MC
CAAAJ61583-MBEA 195.92 CAD
CAAAJ61583-MB CAAAJ61583-MC
Clozapine 97%
Clozapine

Potent, selective muscarinic antagonist


Solid


Applications: Potent, selective muscarinic antagonist

Formula: C₁₈H₁₉ClN₄
Melting Pt: 182…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00153785
CAS Number: 5786-21-0
EINECS: 227-313-7
UN: 2811
ADR: 6.1,III
Merck Index: 14,2423

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR