Bradford's reagent for dye, ready-to-use
Supplier: Thermo Scientific Chemicals
|
Danger
|
J61522-AP
J61522-K2
CAAAJ61522-APEA
172.87
CAD
CAAAJ61522-AP
CAAAJ61522-K2
Bradford's reagent for dye, ready-to-use
Bradford's reagent
Suitable for protein concentration determination (100-1500µg)
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Blue
UN No.:UN1805
Applications: Suitable for protein concentration determination (100-1500µg)
Storage Temperature:
Ambient |
UN:
1805 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...