Acrylamide : Bisacrylamide solution_SC (37.5:1) 30%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acrylamide- N,N'-Methylenebisacrylamide, Acrylamide-Bis

J61505-AP J61505-K2
CAAAJ61505-APEA 255.96 CAD
CAAAJ61505-AP CAAAJ61505-K2
Acrylamide : Bisacrylamide solution_SC (37.5:1) 30%
Acrylamide-Bis (32:1)

Liquid

Storage Temperature: Ambient
MDL Number: MFCD00080848
UN: 3426
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR