Soybean oil

Supplier: Thermo Scientific Chemicals

Synonyms: Glycine Soja (Soybean) Oil, Sojae Oleum

J61399-AP J61399-K2
CAAAJ61399-APEA 71.1 CAD
CAAAJ61399-AP CAAAJ61399-K2
Soybean oil
Soybean oil

Oil


CAS No.: 8001-22-7 Flash Point: 113°(235°F) Physical Form: Oil Density: 0.92 Refractive Index: 1.4743 Appearance: Colorless to yellow MDL No.: MFCD00132356

Density: 0,92
Flash Pt: >113 °C (235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00132356
CAS Number: 8001-22-7
EINECS: 232-274-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR