Nicardipine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-(2-(Benzyl(methyl)amino)ethyl) 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate hydrochloride, Nicardipine HCl

J61269-03 J61269-06
CAAAJ61269-03EA 137.46 CAD
CAAAJ61269-03 CAAAJ61269-06
Nicardipine hydrochloride 98%
Nicardipine hydrochloride

L-type calcium channel blocker


Powder


Applications: L-type calcium channel blocker

Formula: C₂₆H₂₉N₃O₆
MW: 515.99 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00057327
CAS Number: 54527-84-3
EINECS: 259-198-4
UN: 2811
ADR: "6.1",III
Merck Index: 14,6495

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR