Brucine
Supplier: Thermo Scientific Chemicals
|
Danger
|
J61178-06
J61178-14
J61178-22
CAAAJ61178-06EA
44.56
CAD
CAAAJ61178-06
CAAAJ61178-14
CAAAJ61178-22
Brucine
Brucine
Powder
CAS No.: 357-57-3
Molecular Formula: C23H26N2O4
Formula Weight: 394.47
Physical Form: Powder
Appearance: White
Melting Point: 175-178° dec.
Beilstein Reference: 63046
Optical Rotation: -118° (c=2.5 in chloroform)
UN No.: 1570
MDL No.: MFCD00005942
Formula:
C₂₃H₂₆N₂O₄ Melting Pt: 175…178 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005942 CAS Number: 357-57-3 EINECS: 206-614-7 UN: 1570 ADR: 6.1,I Merck Index: 14,1455 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...