Acrylamide : Bisacrylamide solution (19:1) 40%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acrylamide- N,N'-Methylenebisacrylamide, Acrylamide-Bis

J60909-AP J60909-K2
CAAAJ60909-APEA 195.92 CAD
CAAAJ60909-AP CAAAJ60909-K2
Acrylamide : Bisacrylamide solution (19:1) 40%
Acrylamide-Bis (32:1)

Liquid

Storage Temperature: Ambient
MDL Number: MFCD00080848
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR