Aminophylline, anhydrous 98%, powder
Supplier: Thermo Scientific Chemicals
|
Danger
|
J60705-14
J60705-22
CAAAJ60705-14EA
61.3
CAD
CAAAJ60705-14
CAAAJ60705-22
Aminophylline, anhydrous 98%, powder
Aminophylline
A non-selective phosphodiesterase inhibitor
Aminophylline exhibits vasodilatory and anti-inflammatory activity and is occasionally clinically used to treat chronic obstructive pulmonary disorder (COPD).
In animal models of allergen-induced lung inflammation, aminophylline decreases levels of IL-5, IL-8, and eosinophils. It is mostly used to prepare solutions for intravenous injections.
Formula:
C₇H₈N₄O₂ Melting Pt: 269…274 °C Storage Temperature: Freezer |
MDL Number:
MFCD00013221 CAS Number: 317-34-0 EINECS: 206-264-5 UN: 2811 ADR: "6.1",III Merck Index: 14,465 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...