Meloxicam

Supplier: Thermo Scientific Chemicals
Warning

J60635-06 J60635-14
CAAAJ60635-06EA 152.47 CAD
CAAAJ60635-06 CAAAJ60635-14
Meloxicam
Meloxicam

A NSAID with high inhibitory action against cyclooxygenase (COX-2)


Powder


Applications: A NSAID with high inhibitory action against cyclooxygenase (COX-2)

Formula: C₁₄H₁₃N₃O₄S₂
Melting Pt: 255 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00868752
CAS Number: 71125-38-7
ADR: 6.1,III
Merck Index: 14,5826

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR