Manidipine dihydrochloride

Supplier: Thermo Scientific Chemicals
Danger

J60496-MB J60496-MD
CAAAJ60496-MBEA 172.22 CAD
CAAAJ60496-MB CAAAJ60496-MC CAAAJ60496-MD
Manidipine dihydrochloride
Manidipine dihydrochloride

Calcium channel blocker


Crystalline powder


Applications: Calcium channel blocker

Formula: C₃₅H₃₈N₄O₆
Storage Temperature: Ambient
MDL Number: MFCD00896434
CAS Number: 89226-75-5
UN: 2811
ADR: "6.1",III
Merck Index: 14,5743

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR