2-(3-Trifluoromethylanilino)nicotinic acid 99+%

Supplier: Thermo Scientific Chemicals
Warning

J60489-06 J60489-09 J60489-18
CAAAJ60489-06EA 94.96 CAD
CAAAJ60489-06 CAAAJ60489-09 CAAAJ60489-18
2-(3-Trifluoromethylanilino)nicotinic acid 99+%
2-(3-Trifluoromethylanilino)nicotinic acid

Selective cyclooxygenase-2 (COX-2) inhibitor. A NSAID found to be a potent anion channel blocker


Crystalline powder


Applications: Selective cyclooxygenase-2 (COX-2) inhibitor. A NSAID found to be a potent anion channel blocker

Formula: C₁₃H₉F₃N₂O₂
Melting Pt: 203…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00010569
CAS Number: 4394-00-7
EINECS: 224-516-2
ADR: 6.1,III
Merck Index: 14,6531

Order Now

Specification Test Results

Assay Minimum 99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR