Buspirone hydrochloride

Supplier: Thermo Scientific Chemicals
Danger

J60476-03 J60476-06
CAAAJ60476-03EA 96.22 CAD
CAAAJ60476-03 CAAAJ60476-06
Buspirone hydrochloride

A 5-HT-1A serotonin receptor agonist. Affects dopaminergic, serotonergic and noradrenergic pathways


White to off-white powder


Applications: A 5-HT-1A serotonin receptor agonist. Affects dopaminergic, serotonergic and noradrenergic pathways

Formula: C₂₁H₃₂ClN₅O₂
Storage Temperature: Ambient
MDL Number: MFCD00078569
CAS Number: 33386-08-2
EINECS: 251-489-4
UN: 2811
ADR: 6.1,III
Merck Index: 14,1504

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR