Acrylamide : Bisacrylamide solution (39:1) 30%

Supplier: Thermo Scientific Chemicals
Danger

J60412-AP J60412-K2
CAAAJ60412-APEA 189.6 CAD
CAAAJ60412-AP CAAAJ60412-K2
Acrylamide : Bisacrylamide solution (39:1) 30%
Acrylamide-Bis (32:1)

Liquid

Storage Temperature: Ambient
MDL Number: MFCD00080848
UN: 3426
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR