Auramine O ≥80% (dye content)
Supplier: Thermo Scientific Chemicals
|
Danger
|
J60343-14
J60343-18
CAAAJ60343-14EA
79.36
CAD
CAAAJ60343-14
CAAAJ60343-18
Auramine O ≥80% (dye content)
Auramine O
Fluorescent stain for acid-resistant bacteria in sputum.
Powder
Formula:
C₁₇H₂₂ClN₃ Melting Pt: 250 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012484 CAS Number: 2465-27-2 EINECS: 219-567-2 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay Minimum | 80 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...