Sodium mercaptoacetate (sodium thioglycolate) 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
J60290-14
J60290-22
J60290-A1
CAAAJ60290-14EA
51.82
CAD
CAAAJ60290-14
CAAAJ60290-22
CAAAJ60290-A1
Sodium mercaptoacetate (sodium thioglycolate) 98%
Sodium mercaptoacetate (Sodium thioglycolate)
Reagent that protects tryptophan in amino acid analysis
Powder
Formula:
HSCH₂COONa Melting Pt: >300 °C Storage Temperature: Freezer |
MDL Number:
MFCD00043386 CAS Number: 367-51-1 EINECS: 206-696-4 UN: 2923 ADR: "8",III Merck Index: 14,8692 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...