Mecamylamine hydrochloride

Supplier: Thermo Scientific Chemicals
Danger

J60257-MB J60257-MC
CAAAJ60257-MBEA 368.14 CAD
CAAAJ60257-MB CAAAJ60257-MC
Mecamylamine hydrochloride
Mecamylamine hydrochloride

Nicotinyl acetylcholine receptor antagonist


Powder


Applications: Nicotinyl acetylcholine receptor antagonist

Formula: C₁₁H₂₂ClN
Storage Temperature: Ambient
MDL Number: MFCD00151462
CAS Number: 826-39-1
EINECS: 212-555-8
UN: 2811
ADR: "6.1",III
Merck Index: 14,5774

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR