Loperamide hydrochloride ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
J60168-06
J60168-14
CAAAJ60168-06EA
216.46
CAD
CAAAJ60168-06
CAAAJ60168-14
Loperamide hydrochloride ≥98%
Loperamide hydrochloride
At nanomolar concentrations, binds to mu-opioid receptors. L-type calcium channel blocker
Powder
Applications: At nanomolar concentrations, binds to mu-opioid receptors. L-type calcium channel blocker
Formula:
C₂₉H₃₃ClN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00058581 CAS Number: 34552-83-5 EINECS: 252-082-4 UN: 2811 ADR: 6.1,III Merck Index: 14,5571 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...