Acrylamide : Bisacrylamide solution (19:1) 30%

Supplier: Thermo Scientific Chemicals
Danger

J60126-AP J60126-K2
CAAAJ60126-APEA 189.6 CAD
CAAAJ60126-AP CAAAJ60126-K2
Acrylamide : Bisacrylamide solution (19:1) 30%
Acrylamide-Bis (32:1)

Premixed solution for preparing protein PAGE gels


Liquid

Storage Temperature: Ambient
MDL Number: MFCD00080848
UN: 3426
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR