Abamectin 97, powder

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Avermectin B1

J60039-03 J60039-06
CAAAJ60039-03EA 178.54 CAD
CAAAJ60039-03 CAAAJ60039-06
Abamectin 97, powder
Abamectin

Interacts with GABA receptors to cause chloride channel activation


White to yellow powder


Soluble in DMSO

Formula: C₄₈H₇₂O₁₄
MW: 873.10 g/mol
Melting Pt: 150…155 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01769550
CAS Number: 71751-41-2
UN: 2811
ADR: 6.1 III
Merck Index: 14,2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR