MES, 0.5 M Buffer Solutions, pH 9.0, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J62081-AE J62081-AK J62534-AE J62534-AK J62574-AE J62574-AK J63089-AK J63778-AE J63778-AK J60031-AE
CAAAJ61665-AEEA 70.29 CAD
CAAAJ61665-AE CAAAJ61665-AK CAAAJ62081-AE CAAAJ62081-AK CAAAJ62534-AE CAAAJ62534-AK CAAAJ62574-AE CAAAJ62574-AK CAAAJ63089-AE CAAAJ63089-AK CAAAJ63706-AE CAAAJ63706-AK CAAAJ63778-AE CAAAJ63778-AK CAAAJ63944-AE CAAAJ63944-AK CAAAJ60031-AE CAAAJ60031-AK
MES, 0.5 M Buffer Solutions, pH 9.0, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS: 145224-94-8
MDL: MFCD00006181

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR