2-(4-Thiazolyl)benzimidazole 98+% (by HPLC)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Thiabendazole

J60009-09 J60009-22 J60009-36
CAAAJ60009-09EA 68.07 CAD
CAAAJ60009-09 CAAAJ60009-22 CAAAJ60009-36
2-(4-Thiazolyl)benzimidazole 98+% (by HPLC)
2-(4-Thiazolyl)benzimidazole

Fungicide and antihelminthic


  • Used to control mold and fungal growth in plants
  • Used to treat parasitic infections in humans
  • Used clinically to treat threadworm, cutaneous larva migrans, visceral larva migrans and trichinosis


Thiabendazole, a metal chelator and helminth-specific fumarate reductase inhibitor, is used primarily as an anthelmintic to control infections by helminth organisms.


Applications: Fungicide and antihelminthic


Caution: Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Literature References

Formula: C₁₀H₇N₃S
Melting Pt: 304…305 °C
Storage Temperature: Ambient
MDL Number: MFCD00005587
CAS Number: 148-79-8
EINECS: 205-725-8
UN: 3077
ADR: "9",III
Merck Index: 14,9289

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR