Methylsuccinic acid 99%

Supplier: Thermo Scientific Chemicals

H60967-14 H60967-22
CAAAH60967-14EA 165.9 CAD
CAAAH60967-14 CAAAH60967-22
Methylsuccinic acid 99%
Methylsuccinic acid

Purity: 99% CAS No.: 498-21-5 Molecular Formula: C5H8O4 Formula Weight: 132.11 Melting Point: 110-115° Beilstein Reference: 1722946 MDL No.: MFCD00002659

Formula: C₅H₈O₄
Melting Pt: 110…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00002659
CAS Number: 498-21-5
EINECS: 207-857-1

Order Now

Specification Test Results

Assay Minimum 99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR