Formamidine hydrochloride 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Formimidamide hydrochloride

H60947-06 H60947-14
CAAAH60947-06EA 84.85 CAD
CAAAH60947-06 CAAAH60947-14
Formamidine hydrochloride 98%
Formamidine hydrochloride

Purity: 98% CAS No.: 6313-33-3 Molecular Formula: CH4N2.HCl Formula Weight: 80.52 Melting Point: 84-87° Beilstein Reference: 3906935 MDL No.: MFCD00012865

Formula: CH₄N₂
MW: 80.52 g/mol
Melting Pt: 84…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00012865
CAS Number: 6313-33-3
EINECS: 228-639-2

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR