L-Isoserine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-3-Amino-2-hydroxypropanoic acid, L-Isoserine, (S)-3-Amino-2-hydroxypropionic Acid
H60945-03
H60945-MD
CAAAH60945-03EA
189.6
CAD
CAAAH60945-03
CAAAH60945-MD
L-Isoserine 98%
L-Isoserine
Purity: 98%
CAS No.: 632-13-3
Molecular Formula: C3H7NO3
Formula Weight: 105.09
Melting Point: 197-198°
MDL No.: MFCD03428326
Formula:
C₃H₇NO₃ MW: 105.09 g/mol Melting Pt: 197…198 °C Storage Temperature: Ambient |
MDL Number:
MFCD03428326 CAS Number: 632-13-3 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...