1-Pentyl-4-ethynylbenzene 97%

Supplier: Thermo Scientific Chemicals

H60913-04 H60913-09
CAAAH60913-04EA 60.04 CAD
CAAAH60913-04 CAAAH60913-09
1-Pentyl-4-ethynylbenzene 97%
1-Pentyl-4-ethynylbenzene

CAS: 79887-10-8 MDL: MFCD00173884

Formula: C₁₃H₁₆
Boiling Pt: 172 °C
Density: 0.885
Flash Pt: 103 °C(217 °F)
Storage Temperature: Ambient
MDL Number: MFCD00173884
CAS Number: 79887-10-8

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR